Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mttp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163457.1 | NP_001156929.1 | 10 - 11 | 1456 | 90 | |
NM_008642.2 | NP_032668.2 | 8 - 9 | 1146 | 90 | ||
GenBank mRNA | AK136278.1 | - | 10 - 11 | 1456 | 90 | |
AK146553.1 | - | 8 - 9 | 1147 | 90 | ||
BC012686.1 | - | 8 - 9 | 1106 | 4163901 | 90 | |
EU553486.1 | - | 8 - 9 | 1114 | 90 | ||
L47970.1 | - | 8 - 9 | 1093 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |