Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600011019.mm.1 |
TC0600001373.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011019.mm.1 |
NM_008645.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011019.mm.1 |
NM_008645.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011020.mm.1 |
TC0600001373.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011020.mm.1 |
NM_008645.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011020.mm.1 |
NM_008645.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011024.mm.1 |
TC0600001373.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011024.mm.1 |
NM_008645.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011024.mm.1 |
NM_008645.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011022.mm.1 |
TC0600001373.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011022.mm.1 |
NM_008645.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011022.mm.1 |
NM_008645.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011021.mm.1 |
TC0600001373.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011021.mm.1 |
NM_008645.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011021.mm.1 |
NM_008645.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011023.mm.1 |
TC0600001373.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011023.mm.1 |
NM_008645.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011023.mm.1 |
NM_008645.3 |
Refseq |