Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10541434 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541473 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541451 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541454 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541470 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541464 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541466 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541476 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541429 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541439 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541432 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541435 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541440 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541449 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541428 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541438 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541475 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541468 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541427 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541431 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541465 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541478 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541456 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541437 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541452 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541477 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541469 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541447 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541436 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541461 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541433 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541453 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541430 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10541450 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10541446 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10541448 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10541426 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0600011054.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011054.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011054.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011055.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011055.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011055.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011067.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011067.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011067.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011059.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011059.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011059.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011060.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011060.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011060.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011064.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011064.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011064.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011066.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011066.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011066.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011056.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011056.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011056.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011063.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011063.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011063.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011071.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011071.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011071.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011061.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011061.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011061.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011062.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011062.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011062.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011069.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011069.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011069.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011053.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011053.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011053.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011058.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011058.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011058.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011068.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011068.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011068.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011057.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011057.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011057.mm.1 |
NM_008646.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011065.mm.1 |
TC0600001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011065.mm.1 |
NM_008646.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011065.mm.1 |
NM_008646.3 |
Refseq |
Mouse Expression Set 430 |
1448854_s_at |
- |
- |
Mouse Expression Set 430 |
1448854_PM_s_at |
- |
- |
Murine Genome U74v2 Set |
97680_at |
- |
- |
Murine Genome U74v2 Set |
102973_f_at |
- |
- |
Murine Genome U74v2 Set |
97680_PM_at |
- |
- |
Murine Genome U74v2 Set |
102973_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
135778_PM_at |
- |
- |
Murine Genome U74v2 Set |
135778_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1448854_s_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1448854_PM_s_at |
- |
- |