Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008720.2 | NP_032746.2 | 18 - 19 | 2942 | 94 | |
GenBank mRNA | AF003348.1 | - | 18 - 19 | 2919 | 94 | |
AK154282.1 | - | 18 - 19 | 2862 | 94 | ||
AK155379.1 | - | 18 - 19 | 2862 | 94 | ||
BC052437.1 | - | 18 - 19 | 2942 | 6405214 | 94 | |
BC054539.1 | - | 18 - 19 | 2942 | 6405214 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |