Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008738.2 | NP_032764.1 | 1 - 2 | 333 | 72 | |
GenBank mRNA | AK002508.1 | - | 1 - 2 | 473 | 72 | |
BC057993.1 | - | 1 - 2 | 314 | 5345262 | 72 | |
U78109.1 | - | 1 - 2 | 333 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |