Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008738.2 | NP_032764.1 | 2 - 3 | 514 | 125 | |
GenBank mRNA | AK002508.1 | - | 2 - 3 | 654 | 125 | |
BC057993.1 | - | 2 - 3 | 495 | 5345262 | 125 | |
U78109.1 | - | 2 - 3 | 514 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |