Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008858.3 | NP_032884.2 | 10 - 11 | 1658 | 97 | |
GenBank mRNA | Z34524.1 | - | 10 - 11 | 1533 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |