Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ret | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080780.1 | NP_001074249.1 | 10 - 11 | 2494 | 79 | |
NM_009050.2 | NP_033076.2 | 10 - 11 | 2494 | 79 | ||
GenBank mRNA | AF209436.1 | - | 10 - 11 | 1883 | 79 | |
AK044686.1 | - | 10 - 11 | 2081 | 79 | ||
AK051633.1 | - | 10 - 11 | 2073 | 79 | ||
AK084977.1 | - | 10 - 11 | 2144 | 79 | ||
AY326397.1 | - | 10 - 11 | 1883 | 79 | ||
BC059012.1 | - | 10 - 11 | 2468 | 6849696 | 79 | |
X67812.1 | - | 10 - 11 | 1883 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |