Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042779.2 | NP_001036244.1 | 12 - 13 | 1395 | 75 | |
NM_001291537.1 | NP_001278466.1 | 12 - 13 | 1659 | 75 | ||
NM_001291538.1 | NP_001278467.1 | 12 - 13 | 1653 | 75 | ||
NM_001291539.1 | NP_001278468.1 | 11 - 12 | 1336 | 75 | ||
NM_009153.3 | NP_033179.2 | 10 - 11 | 1205 | 75 | ||
NR_111986.1 | 10 - 11 | 1215 | 75 | |||
GenBank mRNA | AK014333.1 | - | 10 - 11 | 1151 | 75 | |
AK033597.1 | - | 11 - 12 | 1346 | 75 | ||
AK037034.1 | - | 12 - 13 | 1384 | 75 | ||
AK043634.1 | - | 11 - 12 | 1289 | 75 | ||
AK048364.1 | - | 11 - 12 | 1617 | 75 | ||
BC090669.1 | - | 11 - 12 | 1316 | 30696359 | 75 | |
BC150758.1 | - | 9 - 10 | 1051 | 9087669 | 75 | |
CN538654.1 | - | 3 - 4 | 251 | 75 | ||
X85990.1 | - | 10 - 11 | 1202 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |