Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081567.1 | NP_001075036.1 | 1 - 2 | 446 | 62 | |
NM_009158.2 | NP_033184.2 | 1 - 2 | 446 | 62 | ||
GenBank mRNA | AK036275.1 | - | 1 - 2 | 371 | 62 | |
AK076990.1 | - | 1 - 2 | 399 | 62 | ||
AK082242.1 | - | 1 - 2 | 449 | 62 | ||
AK163206.1 | - | 1 - 2 | 397 | 62 | ||
BC026697.1 | - | 1 - 2 | 411 | 62 | ||
BC046625.1 | - | 1 - 2 | 385 | 6494163 | 62 | |
L35236.1 | - | 1 - 2 | 189 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |