Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113569.1 | NP_001107041.1 | 4 - 5 | 389 | 74 | |
NM_009295.2 | NP_033321.2 | 4 - 5 | 384 | 74 | ||
GenBank mRNA | AF326545.1 | - | 4 - 5 | 243 | 74 | |
AF326563.1 | - | 4 - 5 | 243 | 74 | ||
BB852672.1 | - | 4 - 5 | 391 | 74 | ||
BC031728.1 | - | 4 - 5 | 316 | 4480978 | 74 | |
D45903.1 | - | 4 - 5 | 278 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |