Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009320.4 | NP_033346.2 | 5 - 6 | 895 | 76 | |
GenBank mRNA | AK143510.1 | - | 5 - 6 | 859 | 76 | |
BC015245.1 | - | 5 - 6 | 832 | 4242193 | 76 | |
L03292.2 | - | 5 - 6 | 867 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |