Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tert | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009354.1 | NP_033380.1 | 4 - 5 | 1950 | 56 | |
GenBank mRNA | AF051911.1 | - | 4 - 5 | 1950 | 56 | |
AF073311.1 | - | 4 - 5 | 1921 | 56 | ||
BC082327.1 | - | 5 - 6 | 1452 | 56 | ||
BC127068.1 | - | 5 - 6 | 1238 | 40110173 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |