Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009370.2 | NP_033396.1 | 1 - 2 | 90 | 85 | |
GenBank mRNA | BC063260.1 | - | 1 - 2 | 90 | 6409765 | 85 |
CT010393.1 | - | 1 - 2 | 83 | 85 | ||
D25540.1 | - | 1 - 2 | 88 | 85 | ||
D28526.1 | - | 1 - 2 | 83 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |