Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfaip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166402.1 | NP_001159874.1 | 2 - 3 | 668 | 68 | |
NM_009397.3 | NP_033423.3 | 3 - 4 | 753 | 68 | ||
GenBank mRNA | AK088297.1 | - | 3 - 4 | 710 | 68 | |
AK151921.1 | - | 3 - 4 | 714 | 68 | ||
AK153238.1 | - | 3 - 4 | 708 | 68 | ||
AK171326.1 | - | 2 - 3 | 667 | 68 | ||
AK172529.1 | - | 2 - 3 | 670 | 68 | ||
BC050259.1 | - | 3 - 4 | 719 | 68 | ||
BC054806.1 | - | 3 - 4 | 753 | 6838303 | 68 | |
BC060221.1 | - | 3 - 4 | 753 | 6843338 | 68 | |
CT010397.1 | - | 2 - 3 | 480 | 68 | ||
U19463.1 | - | 2 - 3 | 480 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |