Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009404.3 | NP_033430.1 | 1 - 2 | 477 | 70 | |
GenBank mRNA | AK155610.1 | - | 1 - 2 | 478 | 70 | |
AK169471.1 | - | 1 - 2 | 450 | 70 | ||
BC138767.1 | - | 1 - 2 | 456 | 8861789 | 70 | |
BC138770.1 | - | 1 - 2 | 456 | 8861792 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |