Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009425.2 | NP_033451.1 | 1 - 2 | 214 | 83 | |
GenBank mRNA | AK142960.1 | - | 1 - 2 | 216 | 83 | |
AK144931.1 | - | 231 | 83 | |||
AK153581.1 | - | 1 - 2 | 264 | 83 | ||
AK157633.1 | - | 215 | 83 | |||
BC011469.1 | - | 1 - 2 | 180 | 83 | ||
BC138807.1 | - | 1 - 2 | 188 | 8861829 | 83 | |
BC138808.1 | - | 1 - 2 | 188 | 8861830 | 83 | |
BY734631.1 | - | 1 - 2 | 231 | 83 | ||
U37522.1 | - | 1 - 2 | 190 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Mouse Chemokine Panel Mouse Tumor Metastasis Panel 1700 Inventoried Probe spans exons |