Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Areg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009704.3 | NP_033834.1 | 4 - 5 | 854 | 84 | |
GenBank mRNA | AK018590.1 | - | 4 - 5 | 870 | 84 | |
AK020273.1 | - | 4 - 5 | 870 | 84 | ||
BC009138.1 | - | 4 - 5 | 854 | 3597695 | 84 | |
CT010320.1 | - | 4 - 5 | 655 | 84 | ||
D12648.1 | - | 4 - 5 | 681 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |