Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110140.3 | NP_001103610.1 | 13 - 14 | 2297 | 64 | |
NM_009722.3 | NP_033852.1 | 13 - 14 | 2297 | 64 | ||
NR_027838.1 | 13 - 14 | 2297 | 64 | |||
GenBank mRNA | AF039893.1 | - | 9 - 10 | 1444 | 64 | |
AJ131821.1 | - | 13 - 14 | 1967 | 64 | ||
AJ223584.1 | - | 13 - 14 | 1967 | 64 | ||
AK220538.1 | - | 13 - 14 | 2246 | 64 | ||
BC054531.1 | - | 13 - 14 | 1956 | 6406916 | 64 | |
BC054748.1 | - | 13 - 14 | 1956 | 6406916 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |