Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009829.3 | NP_033959.1 | 3 - 4 | 846 | 82 | |
GenBank mRNA | AK147345.1 | - | 3 - 4 | 806 | 82 | |
AK160751.1 | - | 3 - 4 | 806 | 82 | ||
AK168954.1 | - | 3 - 4 | 826 | 82 | ||
BC049086.1 | - | 3 - 4 | 825 | 5716186 | 82 | |
CT010207.1 | - | 3 - 4 | 571 | 82 | ||
M83749.1 | - | 3 - 4 | 841 | 82 | ||
M86182.1 | - | 3 - 4 | 735 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |