Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010020.3 | NP_034150.1 | 5 - 6 | 947 | 62 | |
GenBank mRNA | AF109072.1 | - | 5 - 6 | 870 | 62 | |
AF109391.1 | - | 4 - 5 | 795 | 62 | ||
AJ238309.1 | - | 4 - 5 | 803 | 62 | ||
AK032629.1 | - | 5 - 6 | 905 | 62 | ||
AK136885.1 | - | 5 - 6 | 892 | 62 | ||
BC054119.1 | - | 5 - 6 | 947 | 6588897 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |