Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010020.3 | NP_034150.1 | 10 - 11 | 1558 | 60 | |
GenBank mRNA | AF109072.1 | - | 10 - 11 | 1481 | 60 | |
AF109391.1 | - | 9 - 10 | 1406 | 60 | ||
AK032629.1 | - | 10 - 11 | 1516 | 60 | ||
AK136885.1 | - | 10 - 11 | 1503 | 60 | ||
BC054119.1 | - | 10 - 11 | 1558 | 6588897 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |