Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010137.3 | NP_034267.3 | 2 - 3 | 630 | 68 | |
GenBank mRNA | AF045160.1 | - | 2 - 3 | 394 | 68 | |
AK136911.1 | - | 2 - 3 | 631 | 68 | ||
AK141185.1 | - | 2 - 3 | 367 | 68 | ||
AK158430.1 | - | 2 - 3 | 794 | 68 | ||
BC057870.1 | - | 2 - 3 | 352 | 5032291 | 68 | |
D89787.1 | - | 2 - 3 | 592 | 68 | ||
U81983.1 | - | 2 - 3 | 310 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |