Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010184.2 | NP_034314.2 | 1 - 2 | 100 | 66 | |
GenBank mRNA | BC125455.1 | - | 1 - 2 | 65 | 40129970 | 66 |
BC125457.1 | - | 1 - 2 | 64 | 40129972 | 66 | |
CJ142191.1 | - | 1 - 2 | 100 | 66 | ||
J05018.1 | - | 1 - 2 | 85 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |