Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010187.2 | NP_034317.1 | 5 - 6 | 825 | 92 | |
GenBank mRNA | AK089266.1 | - | 5 - 6 | 826 | 92 | |
AK150613.1 | - | 5 - 6 | 826 | 92 | ||
AK170035.1 | - | 5 - 6 | 92 | |||
BC019758.1 | - | 5 - 6 | 92 | |||
BC038070.1 | - | 5 - 6 | 3993915 | 92 | ||
CD773836.1 | - | 2 - 3 | 93 | |||
M14216.1 | - | 5 - 6 | 783 | 92 | ||
U31803.1 | - | 3 - 4 | 640 | 92 | ||
U31804.1 | - | 3 - 4 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |