Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010189.3 | NP_034319.2 | 3 - 4 | 719 | 82 | |
GenBank mRNA | AK146933.1 | - | 3 - 4 | 684 | 82 | |
AK152879.1 | - | 3 - 4 | 655 | 82 | ||
D37874.1 | - | 3 - 4 | 652 | 82 | ||
L17022.1 | - | 2 - 3 | 358 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |