Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010350.2 | NP_034480.2 | 3 - 4 | 661 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mus musculus Probe spans exons |