Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbegf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010415.2 | NP_034545.1 | 2 - 3 | 510 | 63 | |
GenBank mRNA | AK145548.1 | - | 2 - 3 | 479 | 63 | |
AK160575.1 | - | 2 - 3 | 480 | 63 | ||
AK164309.1 | - | 2 - 3 | 511 | 63 | ||
BC089607.1 | - | 2 - 3 | 479 | 6751441 | 63 | |
L07264.1 | - | 2 - 3 | 443 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |