Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifnar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010508.2 | NP_034638.2 | 5 - 6 | 805 | 99 | |
GenBank mRNA | AK132431.1 | - | 5 - 6 | 737 | 99 | |
AK136950.1 | - | 5 - 6 | 781 | 99 | ||
AK141630.1 | - | 5 - 6 | 741 | 99 | ||
AK148246.1 | - | 5 - 6 | 747 | 99 | ||
AK157555.1 | - | 5 - 6 | 772 | 99 | ||
BC043935.1 | - | 5 - 6 | 756 | 4507009 | 99 | |
BC052217.1 | - | 5 - 6 | 765 | 30008497 | 99 | |
BC052429.1 | - | 5 - 6 | 767 | 6836300 | 99 | |
M89641.1 | - | 5 - 6 | 770 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |