Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifnar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010508.2 | NP_034638.2 | 8 - 9 | 1255 | 107 | |
GenBank mRNA | AK132431.1 | - | 8 - 9 | 1187 | 107 | |
AK136950.1 | - | 8 - 9 | 1231 | 107 | ||
AK141630.1 | - | 8 - 9 | 1191 | 107 | ||
AK148246.1 | - | 8 - 9 | 1197 | 107 | ||
AK157555.1 | - | 9 - 10 | 1283 | 107 | ||
BC043935.1 | - | 8 - 9 | 1206 | 4507009 | 107 | |
BC052217.1 | - | 8 - 9 | 1215 | 30008497 | 107 | |
BC052429.1 | - | 8 - 9 | 1217 | 6836300 | 107 | |
M89641.1 | - | 8 - 9 | 1220 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |