Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il6ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010559.2 | NP_034689.2 | 6 - 7 | 1064 | 98 | |
GenBank mRNA | AK046337.1 | - | 6 - 7 | 1064 | 98 | |
AK082270.1 | - | 6 - 7 | 1065 | 98 | ||
BC019388.1 | - | 6 - 7 | 1058 | 98 | ||
X51975.1 | - | 6 - 7 | 955 | 98 | ||
X51976.1 | - | 944 | 98 | |||
X53802.1 | - | 6 - 7 | 1064 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |