Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010568.2 | NP_034698.2 | 17 - 18 | 3826 | 90 | |
GenBank mRNA | J05149.1 | - | 17 - 18 | 3385 | 90 | |
KC356871.1 | - | 18 - 19 | 3373 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |