Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190830.1 | NP_001177759.1 | 13 - 14 | 1988 | 94 | |
NM_010589.6 | NP_034719.2 | 13 - 14 | 1988 | 94 | ||
GenBank mRNA | AK038268.1 | - | 12 - 13 | 1799 | 94 | |
AK088365.1 | - | 13 - 14 | 1837 | 94 | ||
AK142178.1 | - | 13 - 14 | 1826 | 94 | ||
AK156646.1 | - | 13 - 14 | 1810 | 94 | ||
AK164520.1 | - | 13 - 14 | 1828 | 94 | ||
BC105577.1 | - | 12 - 13 | 1791 | 3968386 | 94 | |
BC131646.1 | - | 12 - 13 | 1799 | 40110146 | 94 | |
BC131647.1 | - | 12 - 13 | 1799 | 40110148 | 94 | |
L32955.1 | - | 12 - 13 | 1787 | 94 | ||
L33768.1 | - | 10 - 11 | 2429 | 94 | ||
L40172.1 | - | 13 - 14 | 1849 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |