Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252654.1 | NP_001239583.1 | 11 - 12 | 874 | 89 | |
NM_001252655.1 | NP_001239584.1 | 11 - 12 | 903 | 89 | ||
NM_001252656.1 | NP_001239585.1 | 12 - 13 | 945 | 89 | ||
NM_010598.3 | NP_034728.2 | 12 - 13 | 916 | 89 | ||
GenBank mRNA | AK143166.1 | - | 11 - 12 | 851 | 89 | |
AK152613.1 | - | 12 - 13 | 868 | 89 | ||
AK157053.1 | - | 11 - 12 | 867 | 89 | ||
BC039178.1 | - | 12 - 13 | 846 | 4503373 | 89 | |
L48983.1 | - | 12 - 13 | 908 | 89 | ||
U31908.1 | - | 12 - 13 | 853 | 89 | ||
U65592.1 | - | 11 - 12 | 711 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |