Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010694.1 | NP_034824.1 | 3 - 4 | 298 | 71 | |
GenBank mRNA | BC116816.1 | - | 3 - 4 | 319 | 40126135 | 71 |
BC116818.1 | - | 3 - 4 | 319 | 40126137 | 71 | |
BC144882.1 | - | 3 - 4 | 319 | 71 | ||
D38580.1 | - | 3 - 4 | 298 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |