Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113408.1 | NP_001106879.1 | 7 - 8 | 1007 | 87 | |
NM_010697.1 | NP_034827.1 | 7 - 8 | 1192 | 87 | ||
GenBank mRNA | AB221569.1 | - | 6 - 7 | 549 | 87 | |
AB250383.1 | - | 6 - 7 | 546 | 87 | ||
AF030333.1 | - | 7 - 8 | 710 | 87 | ||
AK048463.1 | - | 2 - 3 | 232 | 87 | ||
BC013624.1 | - | 7 - 8 | 832 | 3708169 | 87 | |
U69270.1 | - | 7 - 8 | 1192 | 87 | ||
U70375.1 | - | 6 - 7 | 542 | 87 | ||
U89488.1 | - | 7 - 8 | 790 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |