Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252658.1 | NP_001239587.1 | 9 - 10 | 1565 | 82 | |
NM_001252659.1 | NP_001239588.1 | 9 - 10 | 1562 | 82 | ||
NM_010700.3 | NP_034830.2 | 9 - 10 | 1565 | 82 | ||
GenBank mRNA | AF425607.1 | - | 9 - 10 | 1413 | 82 | |
AK076243.1 | - | 8 - 9 | 1947 | 82 | ||
AK149950.1 | - | 9 - 10 | 1503 | 82 | ||
AK151968.1 | - | 9 - 10 | 1532 | 82 | ||
AK152103.1 | - | 9 - 10 | 1501 | 82 | ||
AK160006.1 | - | 9 - 10 | 1503 | 82 | ||
AK161104.1 | - | 9 - 10 | 1503 | 82 | ||
AK166466.1 | - | 9 - 10 | 1502 | 82 | ||
AK170264.1 | - | 9 - 10 | 1510 | 82 | ||
BC019207.1 | - | 9 - 10 | 1483 | 5052599 | 82 | |
BC053041.1 | - | 9 - 10 | 1510 | 5704871 | 82 | |
X64414.1 | - | 9 - 10 | 1484 | 82 | ||
Z19521.1 | - | 9 - 10 | 1363 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |