Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285853.1 | NP_001272782.1 | 4 - 5 | 564 | 63 | |
NM_001285854.1 | NP_001272783.1 | 4 - 5 | 561 | 63 | ||
NM_001285855.1 | NP_001272784.1 | 4 - 5 | 629 | 63 | ||
NM_010768.2 | NP_034898.1 | 5 - 6 | 697 | 63 | ||
GenBank mRNA | AK013606.1 | - | 4 - 5 | 378 | 63 | |
AK018182.1 | - | 4 - 5 | 415 | 63 | ||
AK079816.1 | - | 4 - 5 | 357 | 63 | ||
AK158883.1 | - | 4 - 5 | 422 | 63 | ||
BC060086.1 | - | 4 - 5 | 518 | 63 | ||
BC111864.1 | - | 4 - 5 | 375 | 40046807 | 63 | |
D45243.1 | - | 4 - 5 | 629 | 63 | ||
L27738.1 | - | 5 - 6 | 451 | 63 | ||
S77473.1 | - | 4 - 5 | 376 | 63 | ||
U05210.1 | - | 4 - 5 | 350 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |