Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039652.2 | NP_001034741.1 | 1 - 2 | 577 | 72 | |
GenBank mRNA | AB047546.1 | - | 1 - 2 | 492 | 72 | |
AB441734.1 | - | 1 - 2 | 278 | 72 | ||
AB441735.1 | - | 1 - 2 | 278 | 72 | ||
AB441736.1 | - | 1 - 2 | 278 | 72 | ||
AB441737.1 | - | 1 - 2 | 278 | 72 | ||
AB441738.1 | - | 1 - 2 | 278 | 72 | ||
AB441739.1 | - | 1 - 2 | 278 | 72 | ||
AB441740.1 | - | 1 - 2 | 278 | 72 | ||
AB441741.1 | - | 1 - 2 | 278 | 72 | ||
AB441742.1 | - | 1 - 2 | 278 | 72 | ||
AB441743.1 | - | 1 - 2 | 278 | 72 | ||
AF062753.1 | - | 1 - 2 | 344 | 72 | ||
AF074973.1 | - | 1 - 2 | 344 | 72 | ||
AF074974.1 | - | 1 - 2 | 344 | 72 | ||
AF167565.1 | - | 1 - 2 | 344 | 72 | ||
AF167566.1 | - | 1 - 2 | 344 | 72 | ||
AF167567.1 | - | 1 - 2 | 344 | 72 | ||
AF167568.1 | - | 1 - 2 | 344 | 72 | ||
AF260306.1 | - | 2 - 3 | 507 | 72 | ||
AF260307.1 | - | 2 - 3 | 681 | 72 | ||
AF260308.1 | - | 3 - 4 | 636 | 72 | ||
AF346812.1 | - | 1 - 2 | 344 | 72 | ||
AF346813.1 | - | 1 - 2 | 344 | 72 | ||
AF346814.1 | - | 1 - 2 | 344 | 72 | ||
AF400246.1 | - | 2 - 3 | 507 | 72 | ||
AF400247.1 | - | 2 - 3 | 681 | 72 | ||
AF400248.1 | - | 3 - 4 | 636 | 72 | ||
AK038389.1 | - | 1 - 2 | 318 | 72 | ||
AY036621.1 | - | 1 - 2 | 344 | 72 | ||
AY160190.1 | - | 1 - 2 | 344 | 72 | ||
BC119545.1 | - | 1 - 2 | 344 | 40056952 | 72 | |
DQ363376.1 | - | 1 - 2 | 178 | 72 | ||
DQ363377.1 | - | 1 - 2 | 178 | 72 | ||
DQ868787.1 | - | 1 - 2 | 344 | 72 | ||
DQ868788.2 | - | 1 - 2 | 344 | 72 | ||
EF105311.1 | - | 1 - 2 | 344 | 72 | ||
EF105312.1 | - | 1 - 2 | 344 | 72 | ||
EF105313.1 | - | 1 - 2 | 344 | 72 | ||
EF105314.1 | - | 1 - 2 | 344 | 72 | ||
U19380.1 | - | 1 - 2 | 533 | 72 | ||
U26915.1 | - | 1 - 2 | 560 | 72 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus 1700 Probe spans exons |