Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdgfra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083316.1 | NP_001076785.1 | 5 - 6 | 953 | 61 | |
NM_011058.2 | NP_035188.2 | 5 - 6 | 936 | 61 | ||
GenBank mRNA | AK035501.1 | - | 5 - 6 | 893 | 61 | |
AK081664.1 | - | 5 - 6 | 935 | 61 | ||
AK136490.1 | - | 5 - 6 | 922 | 61 | ||
AK139094.1 | - | 4 - 5 | 1168 | 61 | ||
AK145097.1 | - | 5 - 6 | 1536 | 61 | ||
AK147267.1 | - | 5 - 6 | 885 | 61 | ||
AK163952.1 | - | 5 - 6 | 1000 | 61 | ||
BC020405.1 | - | 5 - 6 | 867 | 61 | ||
BC037467.1 | - | 5 - 6 | 875 | 61 | ||
BC053036.1 | - | 5 - 6 | 953 | 5704645 | 61 | |
DQ912857.1 | - | 2 - 3 | 280 | 61 | ||
M84607.1 | - | 5 - 6 | 946 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |