Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3c2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011084.2 | NP_035214.2 | 5 - 6 | 760 | 82 | |
NM_207683.2 | NP_997566.1 | 3506 | 82 | |||
GenBank mRNA | AB008791.1 | - | 3506 | 82 | ||
AB008792.1 | - | 5 - 6 | 755 | 82 | ||
AK054266.1 | - | 5 - 6 | 551 | 82 | ||
AK054271.1 | - | 5 - 6 | 636 | 82 | ||
AK054415.1 | - | 5 - 6 | 703 | 82 | ||
BC061142.1 | - | 5 - 6 | 547 | 82 | ||
BC150813.1 | - | 3498 | 9087724 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |