Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011103.3 | NP_035233.1 | 8 - 9 | 1026 | 69 | |
GenBank mRNA | AB011812.1 | - | 7 - 8 | 789 | 69 | |
AB201453.1 | - | 5 - 6 | 628 | 69 | ||
AB201454.1 | - | 5 - 6 | 628 | 69 | ||
AB201455.1 | - | 5 - 6 | 628 | 69 | ||
AB201456.1 | - | 5 - 6 | 628 | 69 | ||
AF251036.1 | - | 7 - 8 | 789 | 69 | ||
AK144231.1 | - | 8 - 9 | 1031 | 69 | ||
AK171476.1 | - | 8 - 9 | 1028 | 69 | ||
AY545076.1 | - | 7 - 8 | 789 | 69 | ||
BC051416.1 | - | 8 - 9 | 990 | 6515302 | 69 | |
M69042.1 | - | 8 - 9 | 1021 | 69 | ||
X60304.1 | - | 7 - 8 | 802 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |