Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011163.4 | NP_035293.1 | 3 - 4 | 423 | 74 | |
GenBank mRNA | AK028602.1 | - | 3 - 4 | 423 | 74 | |
BC016422.1 | - | 3 - 4 | 386 | 3964935 | 74 | |
M93567.1 | - | 3 - 4 | 365 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |