Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB021291.1 | - | 2 - 3 | 395 | 90 | |
AF038652.1 | - | 1 - 2 | 154 | 90 | ||
AK035993.1 | - | 2 - 3 | 427 | 90 | ||
AK047067.1 | - | 2 - 3 | 411 | 90 | ||
AK143196.1 | - | 2 - 3 | 438 | 90 | ||
AK158292.1 | - | 2 - 3 | 361 | 90 | ||
BC057048.1 | - | 2 - 3 | 397 | 90 | ||
BC090625.1 | - | 2 - 3 | 397 | 90 | ||
BC094567.1 | - | 2 - 3 | 422 | 6834138 | 90 | |
BC139232.1 | - | 2 - 3 | 369 | 8862254 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |