Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161845.2 | NP_001155317.2 | 6 - 7 | 1069 | 105 | |
NM_001161847.2 | NP_001155319.1 | 4 - 5 | 570 | 105 | ||
NM_001161848.2 | NP_001155320.1 | 4 - 5 | 363 | 105 | ||
NM_001161849.2 | NP_001155321.1 | 4 - 5 | 444 | 105 | ||
NM_001161850.2 | NP_001155322.1 | 4 - 5 | 551 | 105 | ||
NM_011361.3 | NP_035491.1 | 4 - 5 | 419 | 105 | ||
GenBank mRNA | AF139638.1 | - | 4 - 5 | 403 | 105 | |
AF205855.1 | - | 4 - 5 | 400 | 105 | ||
AK086892.1 | - | 4 - 5 | 403 | 105 | ||
AK132234.1 | - | 4 - 5 | 491 | 105 | ||
AK146037.1 | - | 4 - 5 | 403 | 105 | ||
AK146062.1 | - | 4 - 5 | 400 | 105 | ||
AK159131.1 | - | 4 - 5 | 403 | 105 | ||
AK167364.1 | - | 4 - 5 | 400 | 105 | ||
BC005720.1 | - | 4 - 5 | 380 | 3594892 | 105 | |
BC070401.1 | - | 6 - 7 | 1069 | 5695396 | 105 | |
BY736002.1 | - | 4 - 5 | 363 | 105 | ||
CK626189.1 | - | 4 - 5 | 551 | 105 | ||
CN709375.1 | - | 4 - 5 | 287 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |