Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011596.5 | NP_035726.2 | 947 | 70 | ||
GenBank mRNA | AB022323.1 | - | 913 | 70 | ||
AF218252.1 | - | 932 | 70 | |||
AK155055.1 | - | 924 | 70 | |||
AK170122.1 | - | 1201 | 70 | |||
BC018369.1 | - | 900 | 70 | |||
BC108991.1 | - | 844 | 40047832 | 70 | ||
BC108992.1 | - | 844 | 40047833 | 70 | ||
BC112905.1 | - | 907 | 3670722 | 70 | ||
M31226.1 | - | 843 | 70 | |||
X55184.1 | - | 844 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |