Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130174.2 | NP_001123646.1 | 10 - 11 | 563 | 81 | |
NM_001130175.2 | NP_001123647.4 | 10 - 11 | 559 | 81 | ||
NM_001130176.1 | NP_001123648.1 | 8 - 9 | 450 | 81 | ||
NM_001130177.2 | NP_001123649.1 | 9 - 10 | 551 | 81 | ||
NM_001130178.2 | NP_001123650.1 | 9 - 10 | 533 | 81 | ||
NM_001130179.2 | NP_001123651.4 | 9 - 10 | 529 | 81 | ||
NM_001130180.2 | NP_001123652.1 | 8 - 9 | 521 | 81 | ||
NM_001130181.2 | NP_001123653.4 | 8 - 9 | 517 | 81 | ||
NM_011619.3 | NP_035749.1 | 8 - 9 | 521 | 81 | ||
GenBank mRNA | AK146945.1 | - | 10 - 11 | 508 | 81 | |
AK163786.1 | - | 9 - 10 | 496 | 81 | ||
AK169061.1 | - | 9 - 10 | 479 | 81 | ||
BC063753.1 | - | 10 - 11 | 539 | 30293959 | 81 | |
BQ554639.1 | - | 9 - 10 | 469 | 81 | ||
L47549.1 | - | 9 - 10 | 447 | 81 | ||
L47550.1 | - | 9 - 10 | 432 | 81 | ||
L47551.1 | - | 8 - 9 | 433 | 81 | ||
L47552.1 | - | 8 - 9 | 442 | 81 | ||
L47553.1 | - | 8 - 9 | 449 | 81 | ||
L47599.1 | - | 9 - 10 | 453 | 81 | ||
L47600.1 | - | 8 - 9 | 450 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |