Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011638.4 | NP_035768.1 | 4 - 5 | 598 | 97 | |
GenBank mRNA | AK088961.1 | - | 4 - 5 | 512 | 97 | |
BC054522.1 | - | 4 - 5 | 483 | 6406097 | 97 | |
M29618.1 | - | 2 - 3 | 364 | 97 | ||
X57349.1 | - | 3 - 4 | 434 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |