Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vegfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185164.1 | NP_001172093.1 | 2 - 3 | 344 | 57 | |
NM_011697.3 | NP_035827.1 | 2 - 3 | 344 | 57 | ||
GenBank mRNA | AK148188.1 | - | 2 - 3 | 343 | 57 | |
BC046303.1 | - | 2 - 3 | 322 | 6492277 | 57 | |
U43836.1 | - | 2 - 3 | 296 | 57 | ||
U43837.1 | - | 2 - 3 | 104 | 57 | ||
U48800.1 | - | 2 - 3 | 198 | 57 | ||
U52820.1 | - | 2 - 3 | 198 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |