Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011755.2 | NP_035885.2 | 1 - 2 | 93 | 79 | |
GenBank mRNA | AK132703.1 | - | 1 - 2 | 96 | 79 | |
AK159080.1 | - | 1 - 2 | 47 | 79 | ||
BB614371.1 | - | 1 - 2 | 93 | 79 | ||
X17617.1 | - | 1 - 2 | 44 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |